Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252043.1 | NP_001238972.1 | 9 | 1492 | |
NM_004783.3 | NP_004774.1 | 9 | 1492 | ||
NM_016151.3 | NP_057235.2 | 9 | 1492 | ||
XM_011545982.1 | XP_011544284.1 | 9 | 1718 | ||
XM_011545983.2 | XP_011544285.1 | 9 | 1718 | ||
XM_011545984.1 | XP_011544286.1 | 9 | 1718 | ||
XM_011545985.1 | XP_011544287.1 | 9 | 1718 | ||
XM_011545986.2 | XP_011544288.1 | 9 | 1718 | ||
GenBank mRNA | AB020688.1 | - | 9 | 1078 | |
AF061943.1 | - | 9 | 1433 | ||
AF263313.1 | - | 8 | 675 | ||
AK291473.1 | - | 9 | 1009 | ||
AL137701.1 | - | 1 | 1329 | ||
BC015204.1 | - | 9 | 1458 | ||
BC031825.2 | - | 9 | 750 | ||
BC051798.1 | - | 9 | 1228 | ||
BC054495.1 | - | 9 | 1445 | ||
BC136653.1 | - | 9 | 1177 | 9020643 | |
BC136655.1 | - | 9 | 1177 | 9020645 | |
BC142663.1 | - | 9 | 832 | 40148112 | |
BC144344.1 | - | 9 | 1177 | 9052864 | |
BC151221.1 | - | 9 | 832 | 40148752 | |
BC152413.1 | - | 9 | 1078 | 8862518 |