Gene Transcripts

Gene Symbol
Lhfpl3
Entrez Gene ID
499977
Gene Name
lipoma HMGIC fusion partner-like 3
Gene Aliases
RGD1559727
Chromosome Location
Chr. 4: 8537590 - 8994097 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762671.1 XP_008760893.1 2 3233

View Full Product Details