Gene Transcripts
- Gene Symbol
- Gars
- Entrez Gene ID
- 297113
- Gene Name
- glycyl-tRNA synthetase
- Gene Aliases
- GlyRS, RGD1559871
- Chromosome Location
- Chr. 4: 85235128 - 85276085 on Build Rnor_6.0
- UniGene ID
- Rn.3338
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC088347.1 |
- |
6, 7 |
584 |
|
View Full Product Details