Gene Transcripts

Gene Symbol
Dcp1b
Entrez Gene ID
500305
Gene Name
decapping mRNA 1B
Gene Aliases
RGD1562214
Chromosome Location
Chr. 4: 151249655 - 151287018 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749821.3 XP_003749869.1 9 2037
XM_008763272.1 XP_008761494.1 9 2205
XM_575654.6 XP_575654.3 9 2136

View Full Product Details