Gene Transcripts

Gene Symbol
B4galnt3
Entrez Gene ID
500306
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 3
Gene Aliases
RGD1561056
Chromosome Location
Chr. 4: 152736060 - 152835488 on Build Rnor_6.0
UniGene ID
Rn.228191
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191976.1 NP_001178905.1 7 683
XM_006237267.2 XP_006237329.1 7 989

View Full Product Details