Gene Transcripts

Gene Symbol
Fam83a
Entrez Gene ID
500873
Gene Name
family with sequence similarity 83, member A
Gene Aliases
RGD1565606
Chromosome Location
Chr. 7: 97853091 - 97880936 on Build Rnor_6.0
UniGene ID
Rn.207265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130576.1 NP_001124048.1 1 456
XM_008765518.1 XP_008763740.1 2 3091

View Full Product Details