Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291824.1 | NP_001278753.1 | 6 | 821 | |
NM_001291825.1 | NP_001278754.1 | 5 | 834 | ||
NM_001291826.1 | NP_001278755.1 | 5 | 729 | ||
NM_001291828.1 | NP_001278757.1 | 3 | 431 | ||
NM_004810.3 | NP_004801.1 | 6 | 807 | ||
XM_006724376.3 | XP_006724439.1 | 5 | 724 | ||
GenBank mRNA | AF042380.1 | - | 6 | 736 | |
AF090456.1 | - | 6 | 698 | ||
AF102694.1 | - | 6 | 736 | ||
AF121002.1 | - | 5 | 841 | ||
AF129476.1 | - | 6 | 726 | ||
AF236119.1 | - | 6 | 767 | ||
AF236120.1 | - | 5 | 544 | ||
AJ011736.1 | - | 6 | 752 | ||
AK303280.1 | - | 5 | 714 | ||
AK303332.1 | - | 4 | 621 | ||
AK303470.1 | - | 3 | 422 | ||
AK303512.1 | - | 3 | 476 | ||
AK303657.1 | - | 5 | 686 | ||
AK312830.1 | - | 6 | 806 | ||
AY069959.1 | - | 6 | 807 | ||
BC025692.1 | - | 6 | 741 | 5208327 | |
BC026002.1 | - | 5 | 803 | 4672797 | |
CR456498.1 | - | 6 | 605 | ||
CR536524.1 | - | 5 | 544 | ||
CR541849.1 | - | 5 | 544 | ||
Y18051.1 | - | 6 | 580 |