Gene Transcripts

Gene Symbol
Ankrd7
Entrez Gene ID
502733
Gene Name
ankyrin repeat domain 7
Gene Aliases
RGD1564660
Chromosome Location
Chr. 4: 45566488 - 45618923 on Build Rnor_6.0
UniGene ID
Rn.228184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001192012.1 NP_001178941.1 3 435
XM_006236119.2 XP_006236181.1 3 1665
XM_006236120.2 XP_006236182.1 3 1668
XM_006236121.2 XP_006236183.1 3 1668

View Full Product Details