Gene Transcripts
- Gene Symbol
- Ldlrad2
- Entrez Gene ID
- 502992
- Gene Name
- low density lipoprotein receptor class A domain containing 2
- Gene Aliases
- RGD1564888
- Chromosome Location
- Chr. 5: 155914496 - 155922269 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_578500.5 |
XP_578500.2
|
2 |
396 |
|
View Full Product Details