Gene Transcripts

Gene Symbol
Cd99
Entrez Gene ID
652929
Gene Name
CD99 molecule
Gene Aliases
Chromosome Location
Chr. 20: 56192426 - 56196961 on Build Rnor_6.0
UniGene ID
Rn.195321
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100804.1 NP_001094274.1 8 450
GenBank mRNA AY262357.1 - 8 439
BC168196.1 - 450 7111460

View Full Product Details