Gene Transcripts

Gene Symbol
Slc6a19
Entrez Gene ID
664630
Gene Name
solute carrier family 6 (neutral amino acid transporter), member 19
Gene Aliases
B0at1
Chromosome Location
Chr. 1: 32199869 - 32218628 on Build Rnor_6.0
UniGene ID
Rn.213283
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039722.2 NP_001034811.1 7, 8 1067
GenBank mRNA AJ890206.1 - 7, 8 1139
EF474455.1 - 7, 8 1100

View Full Product Details