Gene Transcripts

Gene Symbol
Sepsecs
Entrez Gene ID
679383
Gene Name
Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase
Gene Aliases
Chromosome Location
Chr. 14: 60657730 - 60687950 on Build Rnor_6.0
UniGene ID
Rn.216991
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128287.2 NP_001121759.1 9 1084
GenBank mRNA BC166584.1 - 9 1084

View Full Product Details