Gene Transcripts

Gene Symbol
Tirap
Entrez Gene ID
680127
Gene Name
toll-interleukin 1 receptor (TIR) domain-containing adaptor protein
Gene Aliases
Chromosome Location
Chr. 8: 36383907 - 36388249 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001055833.4 XP_001055833.2 2 253
XM_006242794.2 XP_006242856.1 2 253

View Full Product Details