Gene Transcripts

Gene Symbol
Aup1
Entrez Gene ID
680423
Gene Name
ancient ubiquitous protein 1
Gene Aliases
Chromosome Location
Chr. 4: 113887030 - 113890103 on Build Rnor_6.0
UniGene ID
Rn.198163
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079899.1 NP_001073368.1 5 604
XM_008763029.1 XP_008761251.1 5 689
GenBank mRNA BC127544.1 - 5 604
FQ228919.1 - 5 637

View Full Product Details