Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253854.1 | NP_001240783.1 | 12 | 1765 | |
NM_001253855.1 | NP_001240784.1 | 8 | 1496 | ||
NM_012443.3 | NP_036575.1 | 9 | 1410 | ||
NM_172242.2 | NP_758442.1 | 9 | 1410 | ||
XM_005252645.1 | XP_005252702.1 | 9 | 1439 | ||
XM_005252646.1 | XP_005252703.1 | 8 | 1265 | ||
GenBank mRNA | AF079363.1 | - | 9 | 1367 | |
AK289903.1 | - | 9 | 1359 | ||
AK302194.1 | - | 12 | 1740 | ||
AK310247.1 | - | 8 | 1449 | ||
AL080136.1 | - | 9 | 1358 | ||
BC030585.2 | - | 9 | 1358 | 4822996 | |
CR533552.1 | - | 9 | 1243 | ||
HM370395.1 | - | 4 | 421 |