Gene Transcripts

Gene Symbol
Liph
Entrez Gene ID
681694
Gene Name
lipase, member H
Gene Aliases
Chromosome Location
Chr. 11: 82680167 - 82732145 on Build Rnor_6.0
UniGene ID
Rn.200479
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044279.1 NP_001037744.1 7 1377
GenBank mRNA BC062045.1 - 1383
BC107932.1 - 7 1377 7124552

View Full Product Details