Gene Transcripts

Gene Symbol
Slc25a18
Entrez Gene ID
681896
Gene Name
solute carrier family 25 (glutamate carrier), member 18
Gene Aliases
Chromosome Location
Chr. 4: 153330069 - 153350447 on Build Rnor_6.0
UniGene ID
Rn.42043
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044280.2 NP_001037745.2 3 329
GenBank mRNA BC094517.1 - 3 274
CB726923.1 - 3 330

View Full Product Details