Gene Transcripts

Gene Symbol
Treml2
Entrez Gene ID
680844
Gene Name
triggering receptor expressed on myeloid cells-like 2
Gene Aliases
Chromosome Location
Chr. 9: 14659968 - 14669134 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001059150.5 XP_001059150.1 2, 3 1234
XM_006244493.2 XP_006244555.1 2, 3 1235
XM_008766895.1 XP_008765117.1 3, 4 985

View Full Product Details