Gene Transcripts

Gene Symbol
Cyb5rl
Entrez Gene ID
689035
Gene Name
cytochrome b5 reductase-like
Gene Aliases
Chromosome Location
Chr. 5: 126666933 - 126689260 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001069267.4 XP_001069267.2 3 376
XM_006238551.2 XP_006238613.1 3 378
XM_008763922.1 XP_008762144.1 3 365
XM_008763923.1 XP_008762145.1 3 366
XM_008763924.1 XP_008762146.1 3 364
XM_008763925.1 XP_008762147.1 3 352
XM_008763926.1 XP_008762148.1 3 356
XM_008763927.1 XP_008762149.1 3 360
XM_008763928.1 XP_008762150.1 3 377
XM_008763929.1 XP_008762151.1 3 377
XM_008763930.1 XP_008762152.1 3 476

View Full Product Details