Gene Transcripts

Gene Symbol
Ankrd61
Entrez Gene ID
689907
Gene Name
ankyrin repeat domain 61
Gene Aliases
Chromosome Location
Chr. 12: 12756480 - 12760838 on Build Rnor_6.0
UniGene ID
Rn.208808
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044297.1 NP_001037762.1 3 1141
GenBank mRNA BC082099.1 - 3 1141

View Full Product Details