Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014663.2 | NP_055478.2 | 15, 16 | 2493 | |
XM_005271354.3 | XP_005271411.1 | 15, 16 | 2530 | ||
XM_005271355.3 | XP_005271412.1 | 15, 16 | 2448 | ||
XM_005271356.3 | XP_005271413.1 | 16, 17 | 2569 | ||
XM_011542467.2 | XP_011540769.2 | 6, 7 | 1601 | ||
XM_017002912.1 | XP_016858401.1 | 16, 17 | 2638 | ||
GenBank mRNA | AB014577.1 | - | 15, 16 | 2452 | |
AK294596.1 | - | 1, 2 | 78 | ||
BC002558.2 | - | 15, 16 | 2461 | 3138875 |