Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109570.1 | NP_001103040.1 | 4, 5 | 395 | |
XM_006254884.2 | XP_006254946.1 | 4, 5 | 748 | ||
XM_006254885.2 | XP_006254947.1 | 4, 5 | 747 | ||
XM_006254886.2 | XP_006254948.1 | 4, 5 | 748 |