Gene Transcripts

Gene Symbol
Tcl1a
Entrez Gene ID
690575
Gene Name
T-cell leukemia/lymphoma 1A
Gene Aliases
Tcl1
Chromosome Location
Chr. 6: 128888768 - 128894761 on Build Rnor_6.0
UniGene ID
Rn.218634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109601.1 NP_001103071.1 1, 2 176

View Full Product Details