Gene Transcripts

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
Chromosome Location
Chr. 2: 61187455 - 61470714 on Build GRCh38
UniGene ID
Hs.644708
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014709.3 NP_055524.3 80 10350
GenBank mRNA AB011142.2 - 81 10405
AB018272.1 - 23 3279
AJ586138.1 - 78 9875
AK001193.1 - 20 2941
AK022637.1 - 10 1390
AK023171.1 - 12 1627
AL050092.1 - 15 2446
BC022783.1 - 17 2649
BC062325.1 - 8 1104
BC107761.1 - 7 1015

View Full Product Details