Gene Transcripts
- Gene Symbol
- Spx
- Entrez Gene ID
- 691138
- Gene Name
- spexin hormone
- Gene Aliases
- Npq
- Chromosome Location
- Chr. 4: 176629973 - 176635698 on Build Rnor_6.0
- UniGene ID
- Rn.118301
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY383715.1 |
- |
|
332 |
|
View Full Product Details