Gene Transcripts

Gene Symbol
Stk38l
Entrez Gene ID
691337
Gene Name
serine/threonine kinase 38 like
Gene Aliases
RGD1564793
Chromosome Location
Chr. 4: 181027212 - 181089735 on Build Rnor_6.0
UniGene ID
Rn.200326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083336.1 NP_001076805.1 12 1146
XM_008763431.1 XP_008761653.1 11 2043
GenBank mRNA BC160812.1 - 10 1028
EF444939.1 - 12 1146

View Full Product Details