Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008769713.1 | XP_008767935.1 | 2 | 541 | |
XM_008769714.1 | XP_008767936.1 | 2 | 480 | ||
XM_008769715.1 | XP_008767937.1 | 2 | 541 | ||
XM_008769716.1 | XP_008767938.1 | 2 | 2475 | ||
XM_008769717.1 | XP_008767939.1 | 2 | 2453 | ||
XM_008769718.1 | XP_008767940.1 | 2 | 345 | ||
XM_008769719.1 | XP_008767941.1 | 2 | 541 | ||
XM_008769720.1 | XP_008767942.1 | 2 | 541 |