Gene Transcripts
- Gene Symbol
- Wdr59
- Entrez Gene ID
- 690751
- Gene Name
- WD repeat domain 59
- Gene Aliases
- Chromosome Location
- Chr. 19: 43651689 - 43714389 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_008772573.1 |
XP_008770795.1
|
20, 21 |
2227 |
|
XM_008772574.1 |
XP_008770796.1
|
20, 21 |
2227 |
|
XM_008772575.1 |
XP_008770797.1
|
20, 21 |
2227 |
|
XM_008772576.1 |
XP_008770798.1
|
20, 21 |
2207 |
|
XM_008772577.1 |
XP_008770799.1
|
20, 21 |
2207 |
|
XM_008772578.1 |
XP_008770800.1
|
19, 20 |
2169 |
|
XM_008772579.1 |
XP_008770801.1
|
19, 20 |
2149 |
|
XM_008772580.1 |
XP_008770802.1
|
19, 20 |
2149 |
|
XM_008772581.1 |
XP_008770803.1
|
20, 21 |
2227 |
|
XM_008772582.1 |
XP_008770804.1
|
19, 20 |
2149 |
|
View Full Product Details