Gene Transcripts

Gene Symbol
Nxph2
Entrez Gene ID
679932
Gene Name
neurexophilin 2
Gene Aliases
Chromosome Location
Chr. 3: 307225 - 309534 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761617.1 XP_008759839.1 1 667

View Full Product Details