Gene Transcripts
- Gene Symbol
- Odf2l
- Entrez Gene ID
- 685425
- Gene Name
- outer dense fiber of sperm tails 2-like
- Gene Aliases
- RGD1308397
- Chromosome Location
- Chr. 2: 250995024 - 251032908 on Build Rnor_6.0
- UniGene ID
- Rn.139568
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134708.1 |
NP_001128180.1
|
8, 9 |
1019 |
|
XM_006233439.2 |
XP_006233501.1
|
9, 10 |
1459 |
|
XM_006233440.2 |
XP_006233502.1
|
9, 10 |
1603 |
|
XM_006233441.2 |
XP_006233503.1
|
9, 10 |
1587 |
|
XM_006233442.2 |
XP_006233504.1
|
8, 9 |
7447 |
|
XM_006233443.2 |
XP_006233505.1
|
9, 10 |
1591 |
|
XM_006233444.2 |
XP_006233506.1
|
7, 8 |
7367 |
|
XM_006233445.2 |
XP_006233507.1
|
8, 9 |
1511 |
|
XM_006233446.2 |
XP_006233508.1
|
7, 8 |
7314 |
|
XM_006233447.2 |
XP_006233509.1
|
8, 9 |
7441 |
|
XM_006233448.2 |
XP_006233510.1
|
7, 8 |
7366 |
|
XM_006233449.2 |
XP_006233511.1
|
6, 7 |
7237 |
|
XM_008761524.1 |
XP_008759746.1
|
8, 9 |
7443 |
|
XM_008761525.1 |
XP_008759747.1
|
9, 10 |
1731 |
|
GenBank mRNA |
BC166823.1 |
- |
8, 9 |
1019 |
|
View Full Product Details