Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001085404.1 | NP_001078873.1 | 3 | 309 | |
XM_006237453.2 | XP_006237515.1 | 4 | 342 | ||
XM_008763333.1 | XP_008761555.1 | 3 | 1368 | ||
GenBank mRNA | EF100689.1 | - | 3 | 309 | |
FJ404471.1 | - | 3 | 309 |