Gene Transcripts
- Gene Symbol
- Tex12
- Entrez Gene ID
- 690393
- Gene Name
- testis expressed 12
- Gene Aliases
- Chromosome Location
- Chr. 8: 54991504 - 54999447 on Build Rnor_6.0
- UniGene ID
- Rn.159368
- Species
- Rattus norvegicus
- Gene Symbol
- Bco2
- Entrez Gene ID
- 315644
- Gene Name
- beta-carotene oxygenase 2
- Gene Aliases
- Ab2-079, Bcdo2
- Chromosome Location
- Chr. 8: 54965577 - 54990604 on Build Rnor_6.0
- UniGene ID
- Rn.162999
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY325176.1 |
- |
|
64 |
|
View Full Product Details