Gene Transcripts
- Gene Symbol
- AMMECR1
- Entrez Gene ID
- 9949
- Gene Name
- Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
- Gene Aliases
- AMMERC1
- Chromosome Location
- Chr. X: 110194186 - 110440233 on Build GRCh38
- UniGene ID
- Hs.656243
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025580.1 |
NP_001020751.1
|
4, 5 |
855 |
|
NM_001171689.1 |
NP_001165160.1
|
7, 8 |
1153 |
|
NM_015365.2 |
NP_056180.1
|
5, 6 |
966 |
|
GenBank mRNA |
AJ007014.1 |
- |
5, 6 |
1053 |
|
AK001703.1 |
- |
5, 6 |
489 |
|
AK091430.1 |
- |
7, 8 |
1153 |
|
AK314596.1 |
- |
5, 6 |
897 |
|
BC060813.1 |
- |
4, 5 |
788 |
30344824 |
BX648797.1 |
- |
4, 5 |
817 |
|
KU178496.1 |
- |
4, 5 |
774 |
|
KU178497.1 |
- |
5, 6 |
885 |
|
View Full Product Details