Gene Transcripts
- Gene Symbol
- USP3
- Entrez Gene ID
- 9960
- Gene Name
- ubiquitin specific peptidase 3
- Gene Aliases
- SIH003, UBP
- Chromosome Location
- Chr. 15: 63504511 - 63594640 on Build GRCh38
- UniGene ID
- Hs.458499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256702.1 |
NP_001243631.1
|
9, 10 |
1117 |
|
NM_006537.3 |
NP_006528.2
|
10, 11 |
1249 |
|
NR_046341.1 |
|
11, 12 |
1402 |
|
NR_046342.1 |
|
11, 12 |
1365 |
|
XM_017022763.1 |
XP_016878252.1
|
12, 13 |
2248 |
|
XM_017022764.1 |
XP_016878253.1
|
11, 12 |
2184 |
|
XM_017022765.1 |
XP_016878254.1
|
11, 12 |
1390 |
|
XM_017022766.1 |
XP_016878255.1
|
12, 13 |
1433 |
|
XM_017022767.1 |
XP_016878256.1
|
13, 14 |
2364 |
|
XM_017022768.1 |
XP_016878257.1
|
13, 14 |
2438 |
|
GenBank mRNA |
AF073344.1 |
- |
10, 11 |
1116 |
|
AF077040.1 |
- |
9, 10 |
893 |
|
AK022931.1 |
- |
8, 9 |
836 |
|
AK300948.1 |
- |
8, 9 |
907 |
|
AK301236.1 |
- |
9, 10 |
1033 |
|
AK301563.1 |
- |
4, 5 |
548 |
|
AK307810.1 |
- |
10, 11 |
1008 |
|
AK309382.1 |
- |
9, 10 |
1070 |
|
AK314223.1 |
- |
10, 11 |
1168 |
|
AY461579.1 |
- |
11, 12 |
1278 |
|
BC018113.2 |
- |
10, 11 |
1114 |
3916811 |
BC065300.1 |
- |
10, 11 |
1149 |
6146202 |
BC100029.1 |
- |
10, 11 |
1121 |
|
BC107137.1 |
- |
10, 11 |
1109 |
40025968 |
BC107138.1 |
- |
10, 11 |
1109 |
40025969 |
BT007269.1 |
- |
10, 11 |
1020 |
|
BX647760.1 |
- |
11, 12 |
1143 |
|
View Full Product Details