Gene Transcripts
- Gene Symbol
- Prl3d1
- Entrez Gene ID
- 53950
- Gene Name
- Prolactin family 3, subfamily d, member 1
- Gene Aliases
- Csh1, Pl-I, Pl-Im, Pl1, Rhco1, rPl-1
- Chromosome Location
- Chr. 17: 39777255 - 39782877 on Build Rnor_6.0
- UniGene ID
- Rn.112569
- Species
- Rattus norvegicus
- Gene Symbol
- Prl3d2
- Entrez Gene ID
- 691552
- Gene Name
- Prolactin family 3, subfamily d, member 2
- Gene Aliases
- Pl-1
- Chromosome Location
- Chr. 17: 42849469 - 42855494 on Build Rnor_6.0
- UniGene ID
- Rn.198338
- Species
- Rattus norvegicus
View Full Product Details