Gene Transcripts

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
Chromosome Location
Chr. 19: 571277 - 583493 on Build GRCh38
UniGene ID
Hs.501293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322243.1 NP_001309172.1 8 1703
NM_001728.3 NP_001719.2 9 2055
NM_198589.2 NP_940991.1 8 1707
NM_198590.2 NP_940992.1 8 1762
NM_198591.2 NP_940993.1 7 1605
XM_017027173.1 XP_016882662.1 9 1940
GenBank mRNA AK095912.1 - 7 2123
AY358113.1 - 9 1926
BC009040.2 - 8 1570 3867352
D45131.1 - 8 1554
GU557064.1 - 8 1729
GU557065.1 - 7 1572
L10240.1 - 8 1540
M87879.1 - 8 1550
X64364.1 - 8 1583

View Full Product Details