Gene Transcripts

Gene Symbol
MT1M
Entrez Gene ID
4499
Gene Name
metallothionein 1M
Gene Aliases
MT-1M, MT-IM, MT1, MT1K
Chromosome Location
Chr. 16: 56632622 - 56633986 on Build GRCh38
UniGene ID
Hs.647370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176870.2 NP_789846.1 3 368
GenBank mRNA AF136177.1 - 3 345
BC028280.1 - 3 330 5191481
BM918307.1 - 3 386

View Full Product Details