Gene Transcripts

Gene Symbol
PRB4
Entrez Gene ID
5545
Gene Name
proline rich protein BstNI subfamily 4
Gene Aliases
Po
Chromosome Location
Chr. 12: 11307081 - 11310435 on Build GRCh38
UniGene ID
Hs.528651
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261399.2 NP_001248328.1 5 614
NM_002723.5 NP_002714.2 4 821
GenBank mRNA BC035362.1 - 4 870
BC130386.1 - 4 869 40146417

View Full Product Details