Gene Transcripts

Gene Symbol
HIST1H2BI
Entrez Gene ID
8346
Gene Name
histone cluster 1, H2bi
Gene Aliases
H2B/k, H2BFK
Chromosome Location
Chr. 6: 26272976 - 26273412 on Build GRCh38
UniGene ID
Hs.553506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003525.2 NP_003516.1 1 379
GenBank mRNA BC101653.1 - 1 439
BC101655.1 - 1 439 8069161

View Full Product Details