Gene Transcripts
- Gene Symbol
- TOM1L1
- Entrez Gene ID
- 10040
- Gene Name
- target of myb1 like 1 membrane trafficking protein
- Gene Aliases
- SRCASM
- Chromosome Location
- Chr. 17: 54900691 - 54961967 on Build GRCh38
- UniGene ID
- Hs.153504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321173.1 |
NP_001308102.1
|
8, 9 |
1027 |
|
NM_001321174.1 |
NP_001308103.1
|
6, 7 |
863 |
|
NM_001321175.1 |
NP_001308104.1
|
9, 10 |
1035 |
|
NM_001321176.1 |
NP_001308105.1
|
9, 10 |
1030 |
|
NM_005486.2 |
NP_005477.2
|
8, 9 |
1027 |
|
XM_017024002.1 |
XP_016879491.1
|
9, 10 |
947 |
|
GenBank mRNA |
AJ010071.1 |
- |
8, 9 |
882 |
|
AK001893.1 |
- |
8, 9 |
964 |
|
AK223125.1 |
- |
8, 9 |
861 |
|
AK300823.1 |
- |
6, 7 |
684 |
|
AK303912.1 |
- |
7, 8 |
781 |
|
AK303913.1 |
- |
8, 9 |
844 |
|
AK309164.1 |
- |
6, 7 |
722 |
|
AK315039.1 |
- |
8, 9 |
865 |
|
AK315907.1 |
- |
6, 7 |
701 |
|
BC029396.1 |
- |
8, 9 |
861 |
4612157 |
BX110664.1 |
- |
1, 2 |
41 |
|
View Full Product Details