Gene Transcripts

Gene Symbol
KCNG2
Entrez Gene ID
26251
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Gene Aliases
KCNF2, KV6.2
Chromosome Location
Chr. 18: 79797963 - 79901894 on Build GRCh38
UniGene ID
Hs.247905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012283.1 NP_036415.1 2 1226
XM_011525918.2 XP_011524220.1 4 3952
XM_011525920.2 XP_011524222.1 4 1392
XM_017025704.1 XP_016881193.1 3 1791
XM_017025705.1 XP_016881194.1 5 3834
GenBank mRNA AJ011021.1 - 2 1226
AK308622.1 - 2 995

View Full Product Details