Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_037144.1 | 3 | 1501 | ||
GenBank mRNA | AF103731.1 | - | 3 | 1501 | |
AF380162.1 | - | 1 | 1330 | ||
AK024218.1 | - | 1 | 476 | ||
BC028914.1 | - | 1 | 1342 | ||
BC063575.1 | - | 3 | 1382 | 3881751 |