Gene Transcripts
- Gene Symbol
- BAIAP2L2
- Entrez Gene ID
- 80115
- Gene Name
- BAI1 associated protein 2 like 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 38084889 - 38110944 on Build GRCh38
- UniGene ID
- Hs.474822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025045.5 |
NP_079321.3
|
12, 13 |
1606 |
|
XM_005261751.4 |
XP_005261808.1
|
13, 14 |
1607 |
|
XM_011530379.2 |
XP_011528681.1
|
13, 14 |
1681 |
|
XM_011530380.2 |
XP_011528682.1
|
11, 12 |
1509 |
|
XM_011530381.2 |
XP_011528683.1
|
12, 13 |
1500 |
|
XM_011530382.2 |
XP_011528684.1
|
12, 13 |
1888 |
|
XM_011530383.2 |
XP_011528685.1
|
11, 12 |
1826 |
|
XM_011530384.2 |
XP_011528686.1
|
12, 13 |
1764 |
|
XM_011530386.2 |
XP_011528688.1
|
6, 7 |
1225 |
|
XM_011530387.2 |
XP_011528689.1
|
7, 8 |
1010 |
|
XM_011530388.2 |
XP_011528690.1
|
6, 7 |
990 |
|
GenBank mRNA |
AY358164.1 |
- |
12, 13 |
1606 |
|
View Full Product Details