Gene Transcripts

Gene Symbol
NKX6-2
Entrez Gene ID
84504
Gene Name
NK6 homeobox 2
Gene Aliases
GTX, NKX6.2, NKX6B
Chromosome Location
Chr. 10: 132783179 - 132786033 on Build GRCh38
UniGene ID
Hs.134013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177400.2 NP_796374.1 2 582
XM_017016789.1 XP_016872278.1 2 576
GenBank mRNA BC101635.1 - 2 582
BC104823.1 - 2 582 8143826
BF509028.1 - 2 212

View Full Product Details