Gene Transcripts

Gene Symbol
ANKRD9
Entrez Gene ID
122416
Gene Name
ankyrin repeat domain 9
Gene Aliases
Chromosome Location
Chr. 14: 102503649 - 102509791 on Build GRCh38
UniGene ID
Hs.432945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152326.2 NP_689539.1 4 697
XM_005267310.2 XP_005267367.1 3 834
XM_005267311.3 XP_005267368.1 3 466
GenBank mRNA AK291868.1 - 3 363
BC015422.1 - 2 496 4397794
BC027479.1 - 2 214 5114925
BX161460.1 - 4 697

View Full Product Details