Gene Transcripts

Gene Symbol
ACSM2A
Entrez Gene ID
123876
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Gene Aliases
A-923A4.1, ACSM2
Chromosome Location
Chr. 16: 20451461 - 20487669 on Build GRCh38
UniGene ID
Hs.298252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308169.1 NP_001295098.1 13 1781
NM_001308172.1 NP_001295101.1 14 1966
NM_001308954.1 NP_001295883.1 15 2348
XM_017022923.1 XP_016878412.1 15 1941
XM_017022926.1 XP_016878415.1 12 1483
GenBank mRNA AK055867.1 - 15 1762
AK091878.1 - 7 1932
AK096039.1 - 15 2207
AK298359.1 - 13 1670
BC125176.1 - 14 1803 40121949

View Full Product Details