Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318380.1 | NP_001305309.1 | 2, 3 | 250 | |
NM_177533.4 | NP_803877.2 | 2, 3 | 250 | ||
GenBank mRNA | AB087802.1 | - | 2, 3 | 136 | |
AK128196.1 | - | 2, 3 | 239 | ||
AK310449.1 | - | 2, 3 | 227 | ||
BC041584.1 | - | 2, 3 | 212 | 4810207 | |
DA249638.1 | - | 2, 3 | 170 | ||
DA409636.1 | - | 2, 3 | 233 |