Gene Transcripts

Gene Symbol
IFNL2
Entrez Gene ID
282616
Gene Name
interferon lambda 2
Gene Aliases
IL-28A, IL28A
Chromosome Location
Chr. 19: 39268517 - 39270092 on Build GRCh38
UniGene ID
Hs.567792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172138.1 NP_742150.1 6 672
GenBank mRNA AY129148.1 - 6 672
DQ126337.1 - 6 672

View Full Product Details