Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130105.1 | NP_001123577.1 | 16 | 2039 | |
NM_005713.2 | NP_005704.1 | 15 | 1940 | ||
NM_031361.2 | NP_112729.1 | 14 | 1862 | ||
XM_006714513.2 | XP_006714576.1 | 15 | 1940 | ||
XM_011543090.2 | XP_011541392.1 | 15 | 1940 | ||
XM_011543091.2 | XP_011541393.1 | 14 | 1862 | ||
XM_017008919.1 | XP_016864408.1 | 14 | 1862 | ||
GenBank mRNA | AF136450.1 | - | 15 | 1926 | |
AF232930.1 | - | 14 | 1830 | ||
AK000604.1 | - | 10 | 882 | ||
AK091851.1 | - | 15 | 1752 | ||
AK096854.1 | - | 16 | 2196 | ||
AK292087.1 | - | 14 | 1862 | ||
AY453385.1 | - | 14 | 1440 | ||
AY453386.1 | - | 15 | 1518 | ||
BC000102.2 | - | 14 | 1846 | 3505746 |