Gene Transcripts
- Gene Symbol
- ARPC1B
- Entrez Gene ID
- 10095
- Gene Name
- actin related protein 2/3 complex subunit 1B
- Gene Aliases
- ARC41, p40-ARC, p41-ARC
- Chromosome Location
- Chr. 7: 99374675 - 99394801 on Build GRCh38
- UniGene ID
- Hs.489284
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005720.3 |
NP_005711.1
|
4, 5 |
510 |
|
XM_006715825.1 |
XP_006715888.1
|
4, 5 |
521 |
|
XM_006715826.1 |
XP_006715889.1
|
4, 5 |
505 |
|
GenBank mRNA |
AF006084.1 |
- |
4, 5 |
470 |
|
AK314818.1 |
- |
4, 5 |
455 |
|
AU143525.1 |
- |
4, 5 |
510 |
|
AY007111.1 |
- |
2, 3 |
340 |
|
BC002562.2 |
- |
4, 5 |
443 |
3138319 |
BC002988.1 |
- |
4, 5 |
479 |
2821335 |
BC007555.1 |
- |
4, 5 |
441 |
2967684 |
View Full Product Details